missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Triaza-7-phosphaadamantane, 97+%, Thermo Scientific Chemicals

Product Code. 15452829
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53597-69-6
Molecular Formula C6H12N3P
Molecular Weight (g/mol) 157.157
MDL Number MFCD00154905
InChI Key FXXRPTKTLVHPAR-UHFFFAOYSA-N
Synonym 1,3,5-triaza-7-phosphaadamantane, 1,3,5-triaza-7-phosphatricyclo 3.3.1.13,7 decane, 1,3,5-triaza-7-phosphaadamantane,min, acmc-20ap5q, 1,5-triaza-9-phospha-adamantane, 1,5-triaza-7-phosphatricyclo 3.3.1.13,7 decane, 1,3,5-triaza-7-phosphatricyclo 3.3.1.13.7 decane, 1,3,5-triaza-7-phosphatricyclo 3.3.1.1 3,7 decane
PubChem CID 143061
SMILES C1N2CN3CN1CP(C2)C3

Specifications

Melting Point 244°C to 250°C
Color White
Quantity 1 g
Sensitivity Hygroscopic
Formula Weight 157.15
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material 1,3,5-Triaza-7-phosphaadamantane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,3,5-Triaza-7-phosphaadamantane, 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.