Learn More
1,3-acetonedicarboxylic acid, 96%, Thermo Scientific Chemicals
64.32 € - 717.00 €
Chemical Identifiers
CAS | 542-05-2 |
---|---|
Molecular Formula | C5H6O5 |
Molecular Weight (g/mol) | 146.1 |
MDL Number | MFCD00002711 |
InChI Key | OXTNCQMOKLOUAM-UHFFFAOYSA-N |
Synonym | 1,3-acetonedicarboxylic acid, 3-oxoglutaric acid, acetonedicarboxylic acid, pentanedioic acid, 3-oxo, acetone-1,3-dicarboxylic acid, 3-ketoglutaric acid, unii-ih7p7wo21p, 1,3-acetonedicarboxylic acid; 3-ketoglutaric acid, ih7p7wo21p, acetondicarbonsaure |
PubChem CID | 68328 |
IUPAC Name | 3-oxopentanedioic acid |
SMILES | C(C(=O)CC(=O)O)C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10733371
|
Thermo Scientific Acros
173151000 |
100 g | Glass Bottle |
64.32 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10118133
|
Thermo Scientific Acros
173155000 |
500 g | Glass Bottle |
243.21 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15139213
|
Thermo Scientific Acros
173150025 |
2.5 kg | Plastic drum | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
542-05-2 | |
146.1 | |
OXTNCQMOKLOUAM-UHFFFAOYSA-N | |
68328 | |
C(C(=O)CC(=O)O)C(=O)O |
C5H6O5 | |
MFCD00002711 | |
1,3-acetonedicarboxylic acid, 3-oxoglutaric acid, acetonedicarboxylic acid, pentanedioic acid, 3-oxo, acetone-1,3-dicarboxylic acid, 3-ketoglutaric acid, unii-ih7p7wo21p, 1,3-acetonedicarboxylic acid; 3-ketoglutaric acid, ih7p7wo21p, acetondicarbonsaure | |
3-oxopentanedioic acid |
Specifications
542-05-2 | |
100.0 | |
White | |
Authentic | |
C5H6O5 | |
MFCD00002711 | |
11,2 | |
1,3-acetonedicarboxylic acid, 3-oxoglutaric acid, acetonedicarboxylic acid, pentanedioic acid, 3-oxo, acetone-1,3-dicarboxylic acid, 3-ketoglutaric acid, unii-ih7p7wo21p, 1,3-acetonedicarboxylic acid; 3-ketoglutaric acid, ih7p7wo21p, acetondicarbonsaure | |
OXTNCQMOKLOUAM-UHFFFAOYSA-N | |
3-oxopentanedioic acid | |
68328 | |
96% | |
1, 3-Acetonedicarboxylic acid |
95.0 | |
133°C | |
>110°C | |
96% | |
HO2CCH2COCH2CO2H | |
03,789 | |
15,67 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol,slightly soluble in ethyl acetate and ether,insoluble in benzene,chloroform,ligroin,decomposes in hot water,acids,alkalies to co2 an,d acetone | |
C(C(=O)CC(=O)O)C(=O)O | |
146.1 | |
146.1 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective cloth
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 208-797-9
Recommended Storage : preferably stored under cool conditions
- 1,3-Acetonedicarboxylic acid
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P362-Take off contaminated clothing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.