missing translation for 'onlineSavingsMsg'
Learn More
1,3-Bis(1-adamantyl)imidazolium tetrafluoroborate, Thermo Scientific™ Product Code.: 15457647

1,3-Bis(1-adamantyl)imidazolium tetrafluoroborate, Thermo Scientific™

Product Code. 15457647
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15457647

Brand: Thermo Scientific Alfa Aesar H36330.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 286014-42-4
Molecular Formula C23H33BF4N2
Molecular Weight (g/mol) 424.335
MDL Number MFCD04973311
InChI Key KVWCCJYLKCSVME-UHFFFAOYSA-N
Synonym 1,3-bis 1-adamantyl imidazolium tetrafluoroborate, 1,3-di 1-adamantyl imidazolium tetrafluoroborate, 1,3-di adamantan-1-yl-1h-imidazol-3-ium tetrafluoroborate, 1,3-bis adamantan-1-yl-1??-imidazol-1-ylium tetrafluoroborate, iad.hbf4, acmc-209h3n, 1,3-diadamantyl-imidazolium tetrafluoroborate, 1,3-di 1-adamantyl imidazoliumtetrafluoroborate, 1,3-bis tricyclo 3.3.1.13,7 dec-1-yl-1h-imidazolium tetrafluoroborate
PubChem CID 16218105
IUPAC Name 1,3-bis(1-adamantyl)imidazol-1-ium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.C1C2CC3CC1CC(C2)(C3)N4C=C[N+](=C4)C56CC7CC(C5)CC(C7)C6

Specifications

Melting Point 280°C to 286°C
Color Brown to White
Quantity 1 g
Formula Weight 424.33
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 1,3-Bis(1-adamantyl)imidazolium tetrafluoroborate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,3-Bis(1-adamantyl)imidazolium tetrafluoroborate, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.