missing translation for 'onlineSavingsMsg'
Learn More
1,3-Bis(2,6-diisopropylphenyl)imidazolidinium tetrafluoroborate, 97+%, Thermo Scientific™ Product Code.: 10045173

1,3-Bis(2,6-diisopropylphenyl)imidazolidinium tetrafluoroborate, 97+%, Thermo Scientific™

Product Code. 10045173
5 g, Glass bottle
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10045173

Brand: Thermo Scientific Acros 382420050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 282109-83-5
Molecular Formula C27H39BF4N2
Molecular Weight (g/mol) 478.42
MDL Number MFCD04971187
InChI Key KFZBJQUHHALFSR-UHFFFAOYSA-N
Synonym 1,3-bis 2,6-diisopropylphenyl-4,5-dihydroimidazolium tetrafluoroborate, 1,3-bis 2,6-di-i-propylphenyl-4,5-dihydroimidazolium tetrafluoroborate, sipr-hbf4, 1,3-bis 2,6-diisopropylphenyl-4,5-dihydro-1h-imidazol-3-ium tetrafluoroborate, 4,5-dihydro-1,3-bis 2,6-diisopropylphenyl imidazolium tetrafluoroborate, n,n'-bis 2,6-diisopropylphenyl dihydroimidazolium tetrafluoroborate, sipr.hbf4, 1,3-bis-2,6-diisopropylphenyl imidazolinium tetrafluoroborate, n,n'-2,6-diisopropylphenyl dihydroimidazolium tetrafluoroborate, 1,3-bis 2,6-di-i-propylphenyl-4,5-dihydro-imidazolium tetra-fluoroborateShow MoreShow Less
PubChem CID 11375006
IUPAC Name 1,3-bis[2,6-di(propan-2-yl)phenyl]-4,5-dihydroimidazol-1-ium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.CC(C)C1=C(C(=CC=C1)C(C)C)N2CC[N+](=C2)C3=C(C=CC=C3C(C)C)C(C)C

Specifications

CAS Min % 97.0
CAS Max % 100.0
Color White
Elemental Analysis 97.0% min.
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Glass bottle
Quantity 5 g
Formula Weight 478.42
Percent Purity 97+%
Physical Form Powder
Chemical Name or Material 1, 3-Bis(2, 6-diisopropylphenyl)imidazolidinium tetrafluoroborate
Show More Show Less
compliance-icons
Product Identifier
  • 1,3-Bis(2,6-diisopropylphenyl)imidazolidinium tetrafluoroborate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,3-Bis(2,6-diisopropylphenyl)imidazolidinium tetrafluoroborate, 97+%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.