missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis[tris(hydroxymethyl)amino]propane, 99%, Thermo Scientific Chemicals

Product Code. 10526440
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
Packaging:
Glass bottle
Plastic Bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64431-96-5
Molecular Formula C11H26N2O6
Molecular Weight (g/mol) 282.33
MDL Number MFCD00004689
InChI Key HHKZCCWKTZRCCL-UHFFFAOYSA-N
Synonym bis-tris propane, 1,3-bis tris hydroxymethyl methylamino propane, 2,2'-propane-1,3-diylbis azanediyl bis 2-hydroxymethyl propane-1,3-diol, 2-3-2-hydroxy-1,1-dihydroxymethyl-ethylamino-propylamino-2-hydroxymethyl-propane-1,3-diol, 1,3-propanediol, 2,2'-1,3-propanediyldiimino bis 2-hydroxymethyl, 2,2'-propane-1,3-diyldiimino bis 2-hydroxymethyl propane-1,3-diol, 2-3-2-hydroxy-1,1-bis hydroxymethyl ethyl amino propylamino-2-hydroxymethyl propane-1,3-diol, b3p
PubChem CID 125132
ChEBI CHEBI:40947
IUPAC Name 2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol
SMILES C(CNC(CO)(CO)CO)CNC(CO)(CO)CO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 164.0°C to 165.0°C
Color White
Elemental Analysis 46.33 to 47.2% C, 9.82 to 10.02% N
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH2{CH2NHC(CH2OH)3]2
Quantity 100 g
Beilstein 04,III,859
Solubility Information Solubility in water: soluble in cold water
Formula Weight 282.33
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis[tris(hydroxymethyl)amino]propane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'