missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(3-bromopropoxy)-2-nitrobenzene, 90+%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Specifications
Specifications
CAS Min % | 90.0 |
CAS Max % | 100.0 |
Melting Point | 36°C to 39°C |
Color | Yellow |
Boiling Point | 131°C to 132°C (0.5 mmHg) |
Flash Point | 150°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 90% min. (GC) |
Packaging | Glass bottle |
Molecular Formula | C9H10BrNO3 |
Show More |
Safety and Handling
Product Identifier
- 1-(3-Bromopropoxy)-2-nitrobenzene
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'