missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Bromopropoxy)-3-chlorobenzene, 96%, Thermo Scientific™

Product Code. 10506553
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 37142-46-4
Molecular Formula C9H10BrClO
Molecular Weight (g/mol) 249.53
MDL Number MFCD00596672
InChI Key MVKLHPFBHSJVLA-UHFFFAOYSA-N
Synonym 1-3-bromopropoxy-3-chlorobenzene, 3-bromopropyl 3-chlorophenyl ether
PubChem CID 2734929
IUPAC Name 1-(3-bromopropoxy)-3-chlorobenzene
SMILES C1=CC(=CC(=C1)Cl)OCCCBr

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless
Boiling Point 139°C to 148°C (8.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4O(CH2)3Br
Refractive Index 1.5570 to 1.559
Quantity 5 g
Formula Weight 249.53
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1-(3-Bromopropoxy)-3-chlorobenzene
Show More Show Less
compliance-icons
Product Identifier
  • 1-(3-Bromopropoxy)-3-chlorobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.