missing translation for 'onlineSavingsMsg'
Learn More

1,3-Cyclohexanebis(methylamine), 99%, mixture of cis and trans

Product Code. p-96176 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
This item is not returnable. View return policy

Product Code. 10684672

Brand: Thermo Scientific Acros 177480050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2579-20-6 | C8H18N2 | 142.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2579-20-6
Molecular Formula C8H18N2
Molecular Weight (g/mol) 142.24
MDL Number MFCD00001522
InChI Key QLBRROYTTDFLDX-UHFFFAOYSA-N
Synonym 1,3-cyclohexanedimethanamine, 1,3-cyclohexanebis methylamine, 1,3-bis aminomethyl cyclohexane, cyclohexane-1,3-diyldimethanamine, 1,3-di aminomethyl cyclohexane, kodak silver halide solvent hs-103, cyclohexane, 1,3-bis aminomethyl, 3-aminomethyl cyclohexyl methanamine, 3-aminomethyl cyclohexyl methylamine
PubChem CID 17406
IUPAC Name [3-(aminomethyl)cyclohexyl]methanamine
SMILES C1CC(CC(C1)CN)CN

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -70°C
Color Colorless
Density 0.9400g/mL
Boiling Point 220°C
Flash Point 106°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C6H10(CH2NH2)2
Refractive Index 1.4920 to 1.494
Quantity 5 g
Specific Gravity 0.94
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in ethanol,ethyl ether
Formula Weight 142.24
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1, 3-Cyclohexanebis(methylamine), 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,3-Cyclohexanebis(methylamine), mixture of cis and trans
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.