missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Cyclohexanedione, 97%, may cont. up to 1% NaCl, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
50 g
100 g
1000 g
Description
It is used as a nitisinone intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a nitisinone intermediate.
Solubility
Soluble in water (155 g/L) at 20°C, alcohol, chloroform, acetone, boiling benzene, ether (slightly), carbon disulfide (slightly), and petr ether (slightly).
Notes
Air & Light Sensitive. Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. Keep container tightly closed.
It is used as a nitisinone intermediate.
Solubility
Soluble in water (155 g/L) at 20°C, alcohol, chloroform, acetone, boiling benzene, ether (slightly), carbon disulfide (slightly), and petr ether (slightly).
Notes
Air & Light Sensitive. Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. Keep container tightly closed.
Specifications
Specifications
Melting Point | 101°C to 105°C |
Density | 1.1 |
Quantity | 50 g |
Beilstein | 385888 |
Sensitivity | Light sensitive |
Merck Index | 14,3178 |
Solubility Information | Soluble in water (155g/L) at 20°C,alcohol,chloroform,acetone,boiling benzene,ether (slightly),carbon disulfide (slightly),and petr ether (slightly). |
Formula Weight | 112.13 |
Percent Purity | ≥97% |
Concentration or Composition (by Analyte or Components) | May contain up to 1% NaCl |
Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction