missing translation for 'onlineSavingsMsg'
Learn More

1,3-Cyclopentanediol, 95%, mixture of cis and trans, Thermo Scientific Chemicals

Product Code. 10792295
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59719-74-3
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00001367
InChI Key NUUPJBRGQCEZSI-UHFFFAOYSA-N
Synonym 1,3-cyclopentanediol, 1,3-cyclopentanediol, cis, 1,3-cyclopentanediol, mixture of cis and trans, 1,3-cyclopentandiol, 1,4-cyclopentanediol, cyclopentan-1,3-diol, 3-hydroxy cylcopentanol, cyclopentane 1,3-diol, acmc-20ap4n, 1,3-dihydroxycyclopentane
PubChem CID 100165
IUPAC Name cyclopentane-1,3-diol
SMILES C1CC(CC1O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 40.0°C
Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 80.0°C to 85.0°C (0.1 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C5H8(OH)2
Refractive Index 1.483
Quantity 5 g
Beilstein 06, III, 4057
Specific Gravity 1.09
Solubility Information Solubility in water: partially soluble.
Formula Weight 102.13
Percent Purity 95%
Physical Form Liquid After Melting
Chemical Name or Material 1,3-Cyclopentanediol, Mixture of cis and trans
Show More Show Less
Product Identifier
  • 1,3-Cyclopentanediol, mixture of cis and trans

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'