missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diacetoxybenzene, 97%, Thermo Scientific Chemicals

Product Code. 11484573
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-58-7
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00008701
InChI Key STOUHHBZBQBYHH-UHFFFAOYSA-N
Synonym 1,3-diacetoxybenzene, resorcinol diacetate, 1,3-benzenediol, diacetate, m-phenylenediacetate, resorcinol, diacetate, m-phenylene di acetate, 1,3-dihydroxybenzene diacetate, dihydroxybenzene diacetate, 1,3-phenylene diacetate, 1,3-benzenediol, 1,3-diacetate
PubChem CID 7942
IUPAC Name (3-acetyloxyphenyl) acetate
SMILES CC(=O)OC1=CC(=CC=C1)OC(=O)C

Specifications

Density 1.179
Boiling Point 278°C
Flash Point >110°C (230°F)
Refractive Index 1.503
Quantity 50 g
Beilstein 1875007
Formula Weight 194.19
Percent Purity 98%
Chemical Name or Material 1,3-Diacetoxybenzene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'