missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diacetylbenzene, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
1,3-Diacetylbenzene was used in the preparation of polyhydroxylated analogs. It is an important raw material and intermediates in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,3-Diacetylbenzene was used in the preparation of polyhydroxylated analogs. It is an important raw material and intermediates in organic synthesis.
Solubility
Soluble in ethanol, benzene, chloroform.
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
1,3-Diacetylbenzene was used in the preparation of polyhydroxylated analogs. It is an important raw material and intermediates in organic synthesis.
Solubility
Soluble in ethanol, benzene, chloroform.
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Specifications
Specifications
Melting Point | 35°C to 37°C |
Boiling Point | 150°C to 155°C (15 mmHg) |
Flash Point | >110°C (230°F) |
Quantity | 1 g |
Beilstein | 2042511 |
Solubility Information | Soluble in ethanol,benzene,chloroform. |
IUPAC Name | 1-(3-acetylphenyl)ethan-1-one |
Formula Weight | 162.19 |
Percent Purity | 97% |
Chemical Name or Material | 1,3-Diacetylbenzene |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'