missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diaminopropane, 99%, Thermo Scientific Chemicals

22.71 € - 157.79 €

Chemical Identifiers

CAS 109-76-2
Molecular Formula C3H10N2
Molecular Weight (g/mol) 74.13
MDL Number MFCD00008228
InChI Key XFNJVJPLKCPIBV-UHFFFAOYSA-N
Synonym 1,3-Propanediamine, Trimethylenediamine
PubChem CID 5942
ChEBI CHEBI:16841
IUPAC Name propane-1,3-diamine
SMILES NCCCN
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10533434
View Documents
Thermo Scientific Acros
112350500
50 mL Glass bottle
22.71 €
50mL
Please to purchase this item. Need a web account? Register with us today!
10459020
View Documents
Thermo Scientific Acros
112352500
250 mL Glass bottle
57.79 €
250mL
Please to purchase this item. Need a web account? Register with us today!
15770789
View Documents
Thermo Scientific Acros
112350010
1 L Glass bottle
157.79 €
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

109-76-2
74.13
XFNJVJPLKCPIBV-UHFFFAOYSA-N
5942
propane-1,3-diamine
C3H10N2
MFCD00008228
1,3-Propanediamine, Trimethylenediamine
CHEBI:16841
NCCCN

Specifications

109-76-2
100.0
0.8800g/mL
48°C
98.5% min. (GC)
H2N(CH2)3NH2
MFCD00008228
13,160
1,3-Propanediamine, Trimethylenediamine
XFNJVJPLKCPIBV-UHFFFAOYSA-N
propane-1,3-diamine
5942
74.13
Liquid
98.5
-12°C
140°C
Authentic
C3H10N2
1.4565 to 1.4585
04, 261
0.88
Solubility in water: soluble. Other solubilities: miscible with almost all common organic solvents
NCCCN
74.13
CHEBI:16841
99%
1, 3-Diaminopropane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Fatal in contact with skin.
Flammable liquid and vapor.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breat

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rin

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 203-702-7

Product Identifier
  • 1,3-Diaminopropane

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H310-Fatal in contact with skin.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only