missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromo-4-nitrobenzene, 98%, Thermo Scientific™

Product Code. 15448298
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51686-78-3
Molecular Formula C6H3Br2NO2
Molecular Weight (g/mol) 280.903
MDL Number MFCD02660416
InChI Key DXRVYZGVVFZCFP-UHFFFAOYSA-N
PubChem CID 12315646
IUPAC Name 2,4-dibromo-1-nitrobenzene
SMILES C1=CC(=C(C=C1Br)Br)[N+](=O)[O-]

Specifications

Color Yellow
Quantity 250 mg
UN Number UN3459
Synonym 2,4-dibromonitrobenzene, benzene, 2,4-dibromo-1-nitro, 2,4-dibromo-1-nitro-benzene, 1,3-dibromo-4-nitrobenzene, acmc-209kve, 2,4-dibromo-nitrobenzene, ksc594m9r, benzene,2,4-dibromo-1-nitro, 2,4-bis bromanyl-1-nitro-benzene, 2,4-dibromo-1-nitrobenzene;,3-dibromo-4-nitrobenzene
Formula Weight 280.9
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dibromo-4-nitrobenzene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'