missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromo-5,5-dimethylhydantoin, 98%, Thermo Scientific Chemicals

Product Code. 10275500
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-48-5
Molecular Formula C5H6Br2N2O2
Molecular Weight (g/mol) 285.91
MDL Number MFCD00003189
InChI Key VRLDVERQJMEPIF-UHFFFAOYSA-N
Synonym 1,3-dibromo-5,5-dimethylhydantoin, dibromantin, dibromantine, take charge orange, dbdmh, n,n'-dibromodimethylhydantoin, 5,5-dimethyl-1,3-dibromohydantoin, 2,4-imidazolidinedione, 1,3-dibromo-5,5-dimethyl, 1,3-dibromo-5,5-dimethyl-2,4-imidazolidinedione, unii-v9r5f9i7mz
PubChem CID 6479
IUPAC Name 1,3-dibromo-5,5-dimethylimidazolidine-2,4-dione
SMILES CC1(C(=O)N(C(=O)N1Br)Br)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 187.0°C to 191.0°C
Color Cream or Yellow
Flash Point 155°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Iodometry)
Packaging Plastic bottle
Quantity 25 g
Fieser 01,208; 02,108; 12,158; 15,112
Merck Index 15,3022
Solubility Information Solubility in water: 0.1% (25°C). Other solubilities: soluble in chloroform and glacial acetic acid,0.8% in benzene- 0.02% in ccl4 (25°C),insoluble in hexane,decomposes in ethylalcohol and acetone
Formula Weight 285.91
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1, 3-Dibromo-5, 5-dimethylhydantoin, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,3-Dibromo-5,5-dimethylhydantoin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'