missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromoacetone, 95%, Thermo Scientific Chemicals

Product Code. 15407757
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1,3-Dibromoacetone is used as an antibiotic and antimicrobial agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dibromoacetone is used as an antibiotic and antimicrobial agent.

Solubility
Soluble in acetone, chloroform, dichloromethane and methanol.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 816-39-7
Molecular Formula C3H4Br2O
Molecular Weight (g/mol) 215.872
MDL Number MFCD00013540
InChI Key LQQKDSXCDXHLLF-UHFFFAOYSA-N
Synonym 1,3-dibromoacetone, 1,3-dibromo-2-propanone, 2-propanone, 1,3-dibromo, 1,3-dibromopropanone, bromomethyl ketone, bromomethylketone, sgqdllrxbrwsp@, 1,3-dibromacetone, acmc-20ap4e, 2-propanone,3-dibromo
PubChem CID 69952
IUPAC Name 1,3-dibromopropan-2-one
SMILES C(C(=O)CBr)Br

Specifications

Melting Point 29°C to 30°C
Color White to Yellow
Boiling Point 97°C to 98°C (21 mmHg)
Flash Point 102°C (216°F)
Quantity 5 g
UN Number UN1759
Beilstein 1740389
Sensitivity Moisture sensitive
Solubility Information Soluble in acetone,chloroform,dichloromethane and methanol.
Formula Weight 215.87
Percent Purity 98%
Physical Form Crystals or Fused Solid
Chemical Name or Material 1,3-Dibromoacetone
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.