missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dibromoacetone, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
1,3-Dibromoacetone is used as an antibiotic and antimicrobial agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,3-Dibromoacetone is used as an antibiotic and antimicrobial agent.
Solubility
Soluble in acetone, chloroform, dichloromethane and methanol.
Notes
Air and moisture sensitive. Incompatible with oxidizing agents and heat.
1,3-Dibromoacetone is used as an antibiotic and antimicrobial agent.
Solubility
Soluble in acetone, chloroform, dichloromethane and methanol.
Notes
Air and moisture sensitive. Incompatible with oxidizing agents and heat.
Specifications
Specifications
Melting Point | 29°C to 30°C |
Color | White to Yellow |
Boiling Point | 97°C to 98°C (21 mmHg) |
Flash Point | 102°C (216°F) |
Quantity | 1 g |
UN Number | UN1759 |
Beilstein | 1740389 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in acetone,chloroform,dichloromethane and methanol. |
Formula Weight | 215.87 |
Show More |
Safety and Handling
Product Identifier
- 1,3-Dibromoacetone
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : 1,3-Dibromoacetone
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction