missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-2-propanol, 99%

Product Code. 10032320 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
This item is not returnable. View return policy

Product Code. 10032320

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 96-23-1 | C3H6Cl2O | 128.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-23-1
Molecular Formula C3H6Cl2O
Molecular Weight (g/mol) 128.98
MDL Number MFCD00000951
InChI Key DEWLEGDTCGBNGU-UHFFFAOYSA-N
Synonym 1,3-dichloro-2-propanol, dichlorohydrin, 1,3-dichloroisopropanol, 1,3-dichlorohydrin, glycerol 1,3-dichlorohydrin, 2-propanol, 1,3-dichloro, enodrin, 1,3-dichloro-2-hydroxypropane, 1,3-dichloroisopropyl alcohol, sym-glycerol dichlorohydrin
PubChem CID 7289
ChEBI CHEBI:18917
IUPAC Name 1,3-dichloropropan-2-ol
SMILES OC(CCl)CCl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -4.0°C
Color Colorless to Yellow
Density 1.3510g/mL
Boiling Point 172.0°C to 176.0°C
Flash Point 74°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2CH(OH)CH2Cl
Refractive Index 1.4820 to 1.4840
Quantity 250 mL
Beilstein 01, 364
Merck Index 15, 3084
Specific Gravity 1.351
Solubility Information Solubility in water: 13g/L (20°C). Other solubilities: miscible with most common organic solvents
Viscosity 13.9 mPa.s (20°C)
Formula Weight 128.99
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1, 3-Dichloro-2-propanol, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,3-Dichloro-2-propanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.