missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diiodobenzene, 98%, Thermo Scientific Chemicals

Product Code. 10311681
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-00-6
Molecular Formula C6H4I2
Molecular Weight (g/mol) 329.9
MDL Number MFCD00041731
InChI Key SFPQFQUXAJOWNF-UHFFFAOYSA-N
Synonym m-diiodobenzene, benzene, 1,3-diiodo, benzene, m-diiodo, pubchem3833, 1,3-di iodo benzene, 1,3-diiodobenzene, acmc-1b21u, ksc358c5j, 1,3-diiodo-benzen;benzene, 1,3-diiodo-;benzene, m-diiodo-;benzene,1,3-diiodo-;benzene,m-diiodo
PubChem CID 12270
IUPAC Name 1,3-diiodobenzene
SMILES C1=CC(=CC(=C1)I)I

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 33°C to 37°C
Color Yellow to Beige
Density 2.4700g/mL
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H4I2
Quantity 5 g
Specific Gravity 2.47
Solubility Information Solubility in water: insoluble
Formula Weight 329.9
Percent Purity 98%
Physical Form Low Melting Crystalline Mass, Powder or Crystals
Chemical Name or Material 1, 3-Diiodobenzene, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 1,3-Diiodobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'