missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diisopropylbenzene, 96%, Thermo Scientific Chemicals

31.26 € - 57.49 €

Chemical Identifiers

CAS 99-62-7
Molecular Formula C12H18
Molecular Weight (g/mol) 162.276
MDL Number MFCD00008889
InChI Key UNEATYXSUBPPKP-UHFFFAOYSA-N
Synonym 1,3-diisopropylbenzene, m-diisopropylbenzene, benzene, 1,3-bis 1-methylethyl, benzene, m-diisopropyl, m-diisopropylbenzol, 1,3-bis 1-methylethyl benzene, 3-isopropylcumene, unii-482vxw192e, meta-diisopropylbenzene, chembl31352
PubChem CID 7450
IUPAC Name 1,3-di(propan-2-yl)benzene
SMILES CC(C)C1=CC(=CC=C1)C(C)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11466756
SDS View Documents
Thermo Scientific Alfa Aesar
B23904.18
50 g
31.26 €
50g
Please to purchase this item. Need a web account? Register with us today!
11476756
SDS View Documents
Thermo Scientific Alfa Aesar
B23904.30
250 g
57.49 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1,3-Diisopropylbenzene was used in the synthesis of benzene-1,3,5-triphosphonic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diisopropylbenzene was used in the synthesis of benzene-1,3,5-triphosphonic acid.

Solubility
Soluble in water 4.33 mg/L at 25°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

missing translation for 'einecsNumber' : 202-773-1

missing translation for 'rtecsNumber' : CZ6334000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,3-Diisopropylbenzene, 96%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.