missing translation for 'onlineSavingsMsg'
Learn More
1,3-Dimethyl-2-imidazolidinone, 98%, Thermo Scientific Chemicals Product Code.: 11498596

1,3-Dimethyl-2-imidazolidinone, 98%, Thermo Scientific Chemicals

Product Code. 11498596
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11498596

Brand: Thermo Scientific Alfa Aesar A16001.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1,3-Dimethyl-2-imidazolidinone is used as a high-boiling polar aprotic solvent. It is used as a substitute solvent for toxic hexamethylphosphoramide. It finds application in detergents, dyestuffs, electronic materials and in the manufacture of polymers. It is involved in the preparation of 1,2-bis(trimethylsilyl)benzene. Further, it is utilized as a solvent during alfa-regioselective prenylation of imines. In addition to this, it constitutes the mobile phase during size-exclusion chromatographic analysis of cellulose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dimethyl-2-imidazolidinone is used as a high-boiling polar aprotic solvent. It is used as a substitute solvent for toxic hexamethylphosphoramide. It finds application in detergents, dyestuffs, electronic materials and in the manufacture of polymers. It is involved in the preparation of 1,2-bis(trimethylsilyl)benzene. Further, it is utilized as a solvent during alfa-regioselective prenylation of imines. In addition to this, it constitutes the mobile phase during size-exclusion chromatographic analysis of cellulose.

Solubility
Miscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, acid chlorides and phosphorus halides.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 80-73-9
Molecular Formula C5H10N2O
Molecular Weight (g/mol) 114.15
MDL Number MFCD00003188
InChI Key CYSGHNMQYZDMIA-UHFFFAOYSA-N
Synonym 1,3-dimethyl-2-imidazolidinone, n,n'-dimethylethyleneurea, dimethyl imidazolidinone, n,n'-dimethylimidazolidinone, 1,3-dimethylimidazolidone, rhonite 1, dmeu, karbomos tsem, 2-imidazolidinone, 1,3-dimethyl, 1,3-dimethylethyleneureaShow MoreShow Less
PubChem CID 6661
IUPAC Name 1,3-dimethylimidazolidin-2-one
SMILES CN1CCN(C)C1=O

Spécification

Melting Point 8°C
Density 1.058
Boiling Point 224°C to 226°C
Flash Point 93°C (199°F)
Refractive Index 1.472
Quantity 100 g
UN Number UN2810
Beilstein 108808
Sensitivity Hygroscopic
Merck Index 14,3249
Solubility Information Miscible with water.
Formula Weight 114.15
Percent Purity 98%
Chemical Name or Material 1,3-Dimethyl-2-imidazolidinone
Afficher plus Afficher moins

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10254729 Certificate TypeCertificate of Analysis Date11/04/2025 Catalog NumberA16001.14,A16001.36,A16001.22
Lot Number10226043 Certificate TypeCertificate of Analysis Date26/07/2023 Catalog NumberA16001.14,A16001.36,A16001.22
Lot NumberN12F010 Certificate TypeCertificate of Analysis Date30/12/2022 Catalog NumberA16001.14,A16001.36,A16001.22
Lot Number10240801 Certificate TypeCertificate of Analysis Date30/05/2022 Catalog NumberA16001.14,A16001.36,A16001.22
Lot Number10187862 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16001.14,A16001.36,A16001.22
compliance-icons compliance-icons compliance-icons
Identifiant produit
  • 1,3-Dimethyl-2-imidazolidinone
Mention d'avertissement
  • Danger
Catégorie de danger
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
Mention de danger
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H318-Causes serious eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
Conseils de prudence
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Informations supplémentaires
  • MIXTURE LIST-Contains : 1,3-Dimethylimidazolidin-2-one

RUO – Research Use Only

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit
1,3-Dimethyl-2-imidazolidinone, 98%, Thermo Scientific Chemicals >

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis