missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethylnaphthalene, 96%, Thermo Scientific Chemicals

Product Code. 10173191
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 575-41-7
CAS Min % 95.0
CAS Max % 100.0
Melting Point -5°C
Color Yellow
Density 0.9820g/mL
Boiling Point 263°C
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Molecular Formula C12H12
Linear Formula C10H6(CH3)2
Refractive Index 1.6080 to 1.61
MDL Number MFCD00004036
Quantity 1 g
Specific Gravity 0.982
Synonym naphthalene, 1,3-dimethyl, 1,3-dmn, 1,3-dimethyl-naphthalene, acmc-1ajsv, 1,3-dimethyl naphthalene, naphthalene,1,3-dimethyl, 4-05-00-01708 beilstein handbook reference, 1,3-dimethylnaphthalene
Solubility Information Solubility in water: insoluble in water (20°C)
InChI Key QHJMFSMPSZREIF-UHFFFAOYSA-N
SMILES CC1=CC(C)=C2C=CC=CC2=C1
IUPAC Name 1,3-dimethylnaphthalene
Molecular Weight (g/mol) 156.23
PubChem CID 11327
ChEBI CHEBI:48614
Formula Weight 156.23
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1, 3-Dimethylnaphthalene, 96%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'