missing translation for 'onlineSavingsMsg'
Learn More
1,3-Dioxane, 98%, Thermo Scientific Chemicals Product Code.: 15400498

1,3-Dioxane, 98%, Thermo Scientific Chemicals

Product Code. 15400498
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 15400498

Brand: Thermo Scientific Alfa Aesar H54107.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1,3-Dioxane has been employed as solvent in the synthesis of 8- and 9-membered dioxazocines and dioxazonines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dioxane has been employed as solvent in the synthesis of 8- and 9-membered dioxazocines and dioxazonines.

Solubility
Fully miscible in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 505-22-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00006566
InChI Key VDFVNEFVBPFDSB-UHFFFAOYSA-N
Synonym m-dioxane, 1,3-dioxacyclohexane, meta-dioxane, 1,3-propanediol formal, m-dioxin, dihydro, trimethylene glycol methylene ether, dihydro-m-dioxin, unii-b2c8m17i09, ccris 5911, 2,4-dioxane
PubChem CID 10450
ChEBI CHEBI:46924
IUPAC Name 1,3-dioxane
SMILES C1COCOC1

Specifications

Melting Point -45°C
Density 1.032
Boiling Point 105°C to 106°C
Flash Point 5°C (41°F)
Refractive Index 1.418
Quantity 5 g
UN Number UN1165
Beilstein 102532
Solubility Information Fully miscible in water.
Formula Weight 88.11
Percent Purity 98%
Chemical Name or Material 1,3-Dioxane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU01K036 Certificate TypeCertificate of Analysis Date19/08/2024 Catalog NumberH54107.14,H54107.06
Lot NumberM29K012 Certificate TypeCertificate of Analysis Date08/02/2024 Catalog NumberH54107.14,H54107.06
Lot NumberP17J023 Certificate TypeCertificate of Analysis Date10/04/2023 Catalog NumberH54107.14,H54107.06
Lot NumberN02I033 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog NumberH54107.14,H54107.06
Lot NumberY08I004 Certificate TypeCertificate of Analysis Date25/01/2023 Catalog NumberH54107.14,H54107.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,3-Dioxane, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.