missing translation for 'onlineSavingsMsg'
Learn More

(1,3-Dioxolan-2-ylmethyl)magnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Product Code. 11995991
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 180675-22-3, 109-99-9
Molecular Formula C4H7BrMgO2
Molecular Weight (g/mol) 191.31
MDL Number MFCD01321271
InChI Key ZDNTZRAFJZERSL-UHFFFAOYSA-M
Synonym 1,3-dioxolan-2-ylmethyl magnesium bromide solution, magnesium 2-methanidyl-1,3-dioxolane bromide, magnesium 2+ ion 1,3-dioxolan-2-ylmethanide bromide, grignard reagent
PubChem CID 4247254
IUPAC Name magnesium;2-methanidyl-1,3-dioxolane;bromide
SMILES Br[Mg]CC1OCCO1

Specifications

CAS Min % 8.14, 87.79
CAS Max % 12.21, 91.86
Color Brown to White
Density 0.9400g/mL
Boiling Point 65.0°C to 67.0°C
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Quantity 800 mL
Specific Gravity 0.94
Formula Weight 191.31
Physical Form Powder
Chemical Name or Material (1, 3-Dioxolan-2-ylmethyl)magnesium bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • (1,3-Dioxolan-2-ylmethyl)magnesium bromide, 0.5M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'