missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dioxolane, ReagentPlus™, 99%, Honeywell™
Reagent Grade, contains ∼75 ppm BHT as inhibitor, 99%
200.05 € - 481.25 €
Chemical Identifiers
CAS | 646-06-0 |
---|---|
Molecular Formula | C3H6O2 |
Molecular Weight (g/mol) | 74.079 |
MDL Number | MFCD00003207 |
InChI Key | WNXJIVFYUVYPPR-UHFFFAOYSA-N |
Synonym | dioxolane, glycolformal, formal glycol, 1,3-dioxolan, 1,3-dioxacyclopentane, 1,3-dioxalane, ethylene glycol formal, glycol methylene ether, glycol formal, dioxolan czech |
PubChem CID | 12586 |
ChEBI | CHEBI:87597 |
IUPAC Name | 1,3-dioxolane |
SMILES | C1COCO1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15621660
|
Honeywell Research Chemicals
184497-500ML |
500 mL | Glass Bottle | N/A | |||||
15611660
|
Honeywell Research Chemicals
184497-1L |
1 L | Glass Bottle | N/A | |||||
15608330
|
Honeywell Research Chemicals
184497-4L |
4 L | Aluminum Bottle | N/A | |||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
646-06-0 | |
74.079 | |
WNXJIVFYUVYPPR-UHFFFAOYSA-N | |
12586 | |
1,3-dioxolane |
C3H6O2 | |
MFCD00003207 | |
dioxolane, glycolformal, formal glycol, 1,3-dioxolan, 1,3-dioxacyclopentane, 1,3-dioxalane, ethylene glycol formal, glycol methylene ether, glycol formal, dioxolan czech | |
CHEBI:87597 | |
C1COCO1 |
Specifications
646-06-0 | |
Colorless | |
76°C | |
525°F | |
n20/D 1.401(lit.) | |
UN1166 | |
dioxolane, glycolformal, formal glycol, 1,3-dioxolan, 1,3-dioxacyclopentane, 1,3-dioxalane, ethylene glycol formal, glycol methylene ether, glycol formal, dioxolan czech | |
C1COCO1 | |
74.079 | |
CHEBI:87597 | |
70mmHg (20°C) | |
ReagentPlus™ |
-95°C | |
1.06g/mL at 25°C | |
−3°C (26.6°F) | |
C3H6O2 | |
MFCD00003207 | |
102453 | |
WNXJIVFYUVYPPR-UHFFFAOYSA-N | |
1,3-dioxolane | |
12586 | |
74.08g/mol | |
99% | |
1,3-Dioxolane, Contains approx.75 ppm BHT as inhibitor |
Safety and Handling
P210-P305 + P351 + P338-P370 + P378-P403 + P235
H225-H319
missing translation for 'dotInformation' : Class 3 - PG 2 - Dioxolane
missing translation for 'einecsNumber' : 211-463-5
missing translation for 'rtecsNumber' : JH6760000