missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenylacetone, 98+%, Thermo Scientific Chemicals

Product Code. 11387855
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1,3-Diphenylacetone is used in the aldol condensation reaction with benzil (a dicarbonyl) and base to create tetraphenylcyclopentadienone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diphenylacetone is used in the aldol condensation reaction with benzil (a dicarbonyl) and base to create tetraphenylcyclopentadienone.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-04-5
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.28
MDL Number MFCD00004795
InChI Key YFKBXYGUSOXJGS-UHFFFAOYSA-N
Synonym 1,3-diphenylacetone, dibenzyl ketone, 1,3-diphenyl-2-propanone, benzyl ketone, 2-propanone, 1,3-diphenyl, 1,3-diphenylpropanone, alpha,alpha'-diphenylacetone, 1,3-diphenyl-propan-2-one, unii-9y07g5udkq, fema no. 2397
PubChem CID 7593
IUPAC Name 1,3-diphenylpropan-2-one
SMILES O=C(CC1=CC=CC=C1)CC1=CC=CC=C1

Specifications

Melting Point 32°C to 36°C
Density 1.04
Boiling Point 330°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 974767
Solubility Information Slightly soluble in water.
Formula Weight 210.28
Percent Purity ≥98%
Chemical Name or Material 1,3-Diphenylacetone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'