missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenylguanidine, 97%, Thermo Scientific Chemicals

Product Code. 11414230
Click to view available options
Quantity:
100 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1,3-Diphenylguanidine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diphenylguanidine is used as pharmaceutical intermediate.

Solubility
Soluble in alcohol, chloroform, hot benzene, hot toluene, and dilute mineral acids

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-06-7
Molecular Formula C13H13N3
Molecular Weight (g/mol) 211.268
MDL Number MFCD00001758
InChI Key OWRCNXZUPFZXOS-UHFFFAOYSA-N
Synonym 1,3-diphenylguanidine, diphenylguanidine, vulkazit, accelerator d, n,n'-diphenylguanidine, melaniline, denax, vulkacite d, nocceler d, vulkacit d
PubChem CID 7594
IUPAC Name 1,2-diphenylguanidine
SMILES C1=CC=C(C=C1)NC(=NC2=CC=CC=C2)N

Specifications

Melting Point 146°C to 150°C
Density 1.13
Boiling Point >170°C (decomposition)
Flash Point 170°C (338°F)
Odor Characteristic
Quantity 100 g
UN Number UN3077
Beilstein 1875653
Merck Index 14,3324
Solubility Information Soluble in alcohol, chloroform, hot benzene, hot toluene, and dilute mineral acids
Formula Weight 211.27
Percent Purity 97%
Chemical Name or Material 1,3-Diphenylguanidine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'