missing translation for 'onlineSavingsMsg'
Learn More
1,3-Diphenylisobenzofuran, 97%, Thermo Scientific Chemicals Product Code.: 10326980

1,3-Diphenylisobenzofuran, 97%, Thermo Scientific Chemicals

Product Code. 10326980
1 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10326980

Brand: Thermo Scientific Acros 147580010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5471-63-6
Molecular Formula C20H14O
Molecular Weight (g/mol) 270.32
MDL Number MFCD00005931
InChI Key ZKSVYBRJSMBDMV-UHFFFAOYSA-N
Synonym 1,3-diphenylisobenzofuran, diphenylisobenzofuran, isobenzofuran, 1,3-diphenyl, 1,3 diphenylisobenzofuran, 2,5-diphenyl-3,4-benzofuran, acmc-209lj9, 1,3-diphenylbenzo c furan, 1,3-diphenylisobenzo b furan, 1,3-di phenyl-2-benzofuran, #Show MoreShow Less
PubChem CID 21649
IUPAC Name 1,3-diphenyl-2-benzofuran
SMILES C1=CC=C(C=C1)C2=C3C=CC=CC3=C(O2)C4=CC=CC=C4

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 129.0°C to 133.0°C
Color Yellow to Green
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Beilstein 17, 87
Fieser 01,342; 02,178; 04,209
Formula Weight 270.32
Percent Purity 97%
Physical Form Crystalline Solid or Needles
Chemical Name or Material 1, 3-Diphenylisobenzofuran, 99+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461160 Certificate TypeCertificate of Analysis Date26/03/2024 Catalog Number147580050,147580010
Lot NumberA0454045 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number147580050,147580010
Lot NumberA0372856 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number147580050,147580010
Lot NumberA0416304 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number147580050,147580010
Lot NumberA0418379 Certificate TypeCertificate of Analysis Date20/10/2022 Catalog Number147580050,147580010
Product Identifier
  • 1,3-Diphenylisobenzofuran

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,3-Diphenylisobenzofuran, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.