missing translation for 'onlineSavingsMsg'
Learn More

1,3-Indanedione, 97%, Thermo Scientific Chemicals

34.07 € - 297.48 €

Chemical Identifiers

CAS 606-23-5
Molecular Formula C9H6O2
Molecular Weight (g/mol) 146.15
InChI Key UHKAJLSKXBADFT-UHFFFAOYSA-N
Synonym 1,3-indanedione, 1,3-indandione, 1h-indene-1,3 2h-dione, 1,3-diketohydrindene, indan-1,3-dione, 1,3-indanone, indane-1,3-dione, diketohydrindene, 1,3-indandion, unii-4djn7yg35g
PubChem CID 11815
ChEBI CHEBI:78877
IUPAC Name indene-1,3-dione
SMILES C1C(=O)C2=CC=CC=C2C1=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10366850
View Documents Promotion Details
Thermo Scientific Acros
122090050
5 g Glass bottle
34.07 €
5g
Please to purchase this item. Need a web account? Register with us today!
10376850
View Documents Promotion Details
Thermo Scientific Acros
122090250
25 g Glass bottle
102.51 €
25g
Please to purchase this item. Need a web account? Register with us today!
11914851
View Documents Promotion Details
Thermo Scientific Acros
122091000
100 g Glass bottle
297.48 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

606-23-5
146.15
1,3-indanedione, 1,3-indandione, 1h-indene-1,3 2h-dione, 1,3-diketohydrindene, indan-1,3-dione, 1,3-indanone, indane-1,3-dione, diketohydrindene, 1,3-indandion, unii-4djn7yg35g
CHEBI:78877
C1C(=O)C2=CC=CC=C2C1=O
C9H6O2
UHKAJLSKXBADFT-UHFFFAOYSA-N
11815
indene-1,3-dione

Specifications

606-23-5
Beige to White
97%
07,694
UHKAJLSKXBADFT-UHFFFAOYSA-N
indene-1,3-dione
11815
146.15
Powder
129.0°C to 132.0°C
Authentic
C9H6O2
1,3-indanedione, 1,3-indandione, 1h-indene-1,3 2h-dione, 1,3-diketohydrindene, indan-1,3-dione, 1,3-indanone, indane-1,3-dione, diketohydrindene, 1,3-indandion, unii-4djn7yg35g
C1C(=O)C2=CC=CC=C2C1=O
146.15
CHEBI:78877
97%
1, 3-Indanedione
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 210-109-7

Product Identifier
  • 1,3-Indanedione

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only