missing translation for 'onlineSavingsMsg'
Learn More

1-[3-(Trifluoromethoxy)phenyl]ethanol, 97%, Thermo Scientific™

Product Code. 15495107
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 347194-02-9
Molecular Formula C9H9F3O2
Molecular Weight (g/mol) 206.164
MDL Number MFCD06797343
InChI Key BWANKNAGTRWECQ-UHFFFAOYSA-N
Synonym 1-3-trifluoromethoxy phenyl ethanol, 1-3-trifluoromethoxy phenyl ethan-1-ol, 1-3-trifluoromethoxyphenyl ethanol
PubChem CID 44891072
IUPAC Name 1-[3-(trifluoromethoxy)phenyl]ethanol
SMILES CC(C1=CC(=CC=C1)OC(F)(F)F)O

Specifications

Quantity 5 g
Formula Weight 206.16
Percent Purity 97%
Chemical Name or Material 1-[3-(Trifluoromethoxy)phenyl]ethanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'