missing translation for 'onlineSavingsMsg'
Learn More
1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrrolidine, ≥97%, Thermo Scientific™ Product Code.: 10259562

1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrrolidine, ≥97%, Thermo Scientific™

Product Code. 10259562
250 mg, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10259562

Brand: Thermo Scientific Maybridge CC30139CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 852227-90-8
Molecular Formula C16H24BNO2
Molecular Weight (g/mol) 273.18
MDL Number MFCD08060504
InChI Key DWJNNJSONWFVBT-UHFFFAOYSA-N
Synonym 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrrolidine, 4-pyrrolidinophenylboronic acid pinacol ester, 4-1-pyrrolidinyl benzeneboronic acid pinacol ester, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrrolidine, 4-pyrrolidinophenylboronic acid, pinacol ester, 4-pyrrolidin-1-yl phenylboronic acid pinacol ester, 4-pyrrolidin-1-yl benzeneboronic acid, pinacol ester, 4-pyrrolidin-1-yl phenyl boronic acid pinacol ester, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl phenyl pyrrolidine
PubChem CID 18525715
IUPAC Name 1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrrolidine
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)N1CCCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 273.18
Percent Purity 97%
Chemical Name or Material 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrrolidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrrolidine, ≥97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.