Learn More
1,4,8,11-Tetraazacyclotetradecane, 98+%, Thermo Scientific Chemicals
Description
1,4,8,11-Tetraazacyclotetradecane is used in the synthesis of molecules with electroactive cavities. It acts as a nitrogen crown ether analogue and as an antioxidant in rubber. It is also used in the preparation of plerixafor derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,4,8,11-Tetraazacyclotetradecane is used in the synthesis of molecules with electroactive cavities. It acts as a nitrogen crown ether analogue and as an antioxidant in rubber. It is also used in the preparation of plerixafor derivatives.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with oxidizing agents.
Specifications
Specifications
Melting Point | 184°C to 188°C |
Quantity | 25 g |
Beilstein | 111811 |
Sensitivity | Air sensitive; Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 200.33 |
Percent Purity | ≥98% |
Chemical Name or Material | 1,4,8,11-Tetraazacyclotetradecane |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.