missing translation for 'onlineSavingsMsg'
Learn More

1,4,8,11-Tetraazatricyclo[9.3.1.1(4,8)]hexadecane

Product Code. 15493517
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15493517

Brand: Thermo Scientific Alfa Aesar H26674.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 75920-10-4 | C12H24N4 | 224.352 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75920-10-4
Molecular Formula C12H24N4
Molecular Weight (g/mol) 224.352
MDL Number MFCD09953472
InChI Key ZJCWEHQOOSNWTF-UHFFFAOYSA-N
PubChem CID 337907
SMILES C1CN2CCN3CCCN(C3)CCN(C1)C2

Specifications

Melting Point 83°C to 85°C
Quantity 1 g
Sensitivity Hygroscopic
Synonym 1,4,8,11-tetraazatricyclo 9.3.1.1 4,8 hexadecane, 1,4,8,11-tetraazatricyclo 9.3.1.14,8 hexadecane, 1,4,8,11-tetraazatricyclo 9.3.1.1?,? hexadecane, formaldehyde-cyclam, acmc-20ap3h, cyclam formaldehyde derivative, 1,4,8,11-tetraazatricyclohexadecane, 1,4,8,11-tetraazatricyclo 9.3.1.1∼4,8∼ hexadecane
Formula Weight 224.35
Chemical Name or Material 1,4,8,11-Tetraazatricyclo[9.3.1.1(4,8)]hexadecane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.