missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzenedimethanethiol, Thermo Scientific Chemicals

Product Code. 17291316 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 17291316

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-09-9
Molecular Formula C8H10S2
Molecular Weight (g/mol) 170.29
InChI Key IYPNRTQAOXLCQW-UHFFFAOYSA-N
IUPAC Name [4-(sulfanylmethyl)phenyl]methanethiol
SMILES SCC1=CC=C(CS)C=C1

Specifications

Melting Point 45.0°C to 46.0°C
Boiling Point 156.0°C (16.0 hPa)
Quantity 5 g
Formula Weight 170.3
Chemical Name or Material 1,4-Benzenedimethanethiol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.