missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzodioxan-6-amine, 99%, Thermo Scientific Chemicals

Product Code. 10606772
Click to view available options
Quantity:
5 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22013-33-8
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00006824
InChI Key BZKOZYWGZKRTIB-UHFFFAOYSA-N
Synonym 1,4-benzodioxan-6-amine, 3,4-ethylenedioxyaniline, 6-amino-1,4-benzodioxane, 2,3-dihydrobenzo b 1,4 dioxin-6-amine, 6-amino-1,4-benzodioxan, 1,4-benzodioxan-6-ylamine, 1,4-benzodioxin-6-amine, 2,3-dihydro, 3,4-ethylenedioxy aniline, 2,3-dihydro-benzo 1,4 dioxin-6-ylamine, 2,3-dihydro-1,4-benzodioxin-6-amin
PubChem CID 89148
IUPAC Name 2,3-dihydro-1,4-benzodioxin-6-amine
SMILES NC1=CC=C2OCCOC2=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Brown to Black
Density 1.2300g/mL
Flash Point 120°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.5977 to 1.5997
Quantity 50 g
Specific Gravity 1.23
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in many organic solvents
Formula Weight 151.16
Percent Purity 99%
Physical Form Viscous Liquid After Melting
Chemical Name or Material 1, 4-Benzodioxan-6-amine
Show More Show Less
compliance-icons
Product Identifier
  • 1,4-Benzodioxan-6-amine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.