missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Biphenylyl)ethanol, 98%, Thermo Scientific Chemicals

Product Code. 11311947
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3562-73-0
Molecular Formula C14H14O
Molecular Weight (g/mol) 198.265
MDL Number MFCD00016859
InChI Key GOISDOCZKZYADO-UHFFFAOYSA-N
Synonym 1-4-biphenylyl ethanol, 4-1-hydroxyethyl biphenyl, 1-biphenyl-4-yl ethanol, diascleril, difencol, drucoles, 1-4-phenylphenyl ethanol, 1-4-biphenyl ethanol, 4-biphenylmethanol, .alpha.-methyl, 1-1,1'-biphenyl-4-yl ethanol
PubChem CID 96176
IUPAC Name 1-(4-phenylphenyl)ethanol
SMILES CC(C1=CC=C(C=C1)C2=CC=CC=C2)O

Specifications

Melting Point 94°C to 99°C
Quantity 2 g
Beilstein 2044710
Formula Weight 198.27
Percent Purity 98%
Chemical Name or Material 1-(4-Biphenylyl)ethanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'