missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Bromophenyl)-2-hydroxyethan-1-one, Thermo Scientific™

41.90 € - 367.00 €

Chemical Identifiers

CAS 3343-45-1
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00174275
InChI Key FGROGLJVXNYNQC-UHFFFAOYSA-N
Synonym 1-4-bromophenyl-2-hydroxyethanone, 1-4-bromophenyl-2-hydroxyethan-1-one, ethanone,1-4-bromophenyl-2-hydroxy, 1-4-bromophenyl-2-hydroxy-1-ethanone, 1-4-bromophenyl-2-hydroxyethane-1-one, p-bromphenacylalkohol, zlchem 616, bromophenylhydroxyethanone, p-bromo-a-hydroxy acetophenone, 4-bromobenzoyl methanol
PubChem CID 520633
IUPAC Name 1-(4-bromophenyl)-2-hydroxyethanone
SMILES C1=CC(=CC=C1C(=O)CO)Br
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10181082
View Documents
Thermo Scientific Maybridge
TL00596DA
1 g Amber Glass Bottle
41.90 €
1g
Please to purchase this item. Need a web account? Register with us today!
10467442
View Documents
Thermo Scientific Maybridge
TL00596EA
10 g Amber Glass Bottle
367.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

3343-45-1
215.046
FGROGLJVXNYNQC-UHFFFAOYSA-N
520633
C1=CC(=CC=C1C(=O)CO)Br
C8H7BrO2
MFCD00174275
1-4-bromophenyl-2-hydroxyethanone, 1-4-bromophenyl-2-hydroxyethan-1-one, ethanone,1-4-bromophenyl-2-hydroxy, 1-4-bromophenyl-2-hydroxy-1-ethanone, 1-4-bromophenyl-2-hydroxyethane-1-one, p-bromphenacylalkohol, zlchem 616, bromophenylhydroxyethanone, p-bromo-a-hydroxy acetophenone, 4-bromobenzoyl methanol
1-(4-bromophenyl)-2-hydroxyethanone

Specifications

3343-45-1
97%
BrC6H4C(O)CH2OH
1-4-bromophenyl-2-hydroxyethanone, 1-4-bromophenyl-2-hydroxyethan-1-one, ethanone,1-4-bromophenyl-2-hydroxy, 1-4-bromophenyl-2-hydroxy-1-ethanone, 1-4-bromophenyl-2-hydroxyethane-1-one, p-bromphenacylalkohol, zlchem 616, bromophenylhydroxyethanone, p-bromo-a-hydroxy acetophenone, 4-bromobenzoyl methanol
C1=CC(=CC=C1C(=O)CO)Br
215.046
215.05
1-(4-Bromophenyl)-2-hydroxyethan-1-one
97.0
C8H7BrO2
MFCD00174275
FGROGLJVXNYNQC-UHFFFAOYSA-N
1-(4-bromophenyl)-2-hydroxyethanone
520633
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.