missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Bromophenyl)hexane, 97%, Thermo Scientific™

12.90 € - 166.00 €

Chemical Identifiers

CAS 23703-22-2
Molecular Formula C12H17Br
Molecular Weight (g/mol) 241.17
MDL Number MFCD00061114
InChI Key MXHOLIARBWJKCR-UHFFFAOYSA-N
Synonym 1-4-bromophenyl hexane, 4-hexylbromobenzene, 1-bromo-4-n-hexylbenzene, 4-bromo-1-hexylbenzene, benzene, 1-bromo-4-hexyl, 4-bromo-n-hexylbenzene, 4-hexylphenyl bromide, 1-bromo-4-hexyl-benzene, acmc-1cb1g, #
PubChem CID 598186
IUPAC Name 1-bromo-4-hexylbenzene
SMILES CCCCCCC1=CC=C(Br)C=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10481484
View Documents
Thermo Scientific Maybridge
BTB09866DA
1 g
12.90 €
1g
Please to purchase this item. Need a web account? Register with us today!
10061464
View Documents
Thermo Scientific Maybridge
BTB09866EA
10 g
75.70 €
10g
Please to purchase this item. Need a web account? Register with us today!
10631036
View Documents
Thermo Scientific Maybridge
BTB09866EB
25 g
166.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

23703-22-2
241.17
MXHOLIARBWJKCR-UHFFFAOYSA-N
598186
CCCCCCC1=CC=C(Br)C=C1
C12H17Br
MFCD00061114
1-4-bromophenyl hexane, 4-hexylbromobenzene, 1-bromo-4-n-hexylbenzene, 4-bromo-1-hexylbenzene, benzene, 1-bromo-4-hexyl, 4-bromo-n-hexylbenzene, 4-hexylphenyl bromide, 1-bromo-4-hexyl-benzene, acmc-1cb1g, #
1-bromo-4-hexylbenzene

Specifications

23703-22-2
100.0
Amber Glass Bottle
BrC6H4(CH2)5CH3
1-4-bromophenyl hexane, 4-hexylbromobenzene, 1-bromo-4-n-hexylbenzene, 4-bromo-1-hexylbenzene, benzene, 1-bromo-4-hexyl, 4-bromo-n-hexylbenzene, 4-hexylphenyl bromide, 1-bromo-4-hexyl-benzene, acmc-1cb1g, #
CCCCCCC1=CC=C(Br)C=C1
241.17
241.17
1-(4-Bromophenyl)hexane
96.5
97%
C12H17Br
MFCD00061114
MXHOLIARBWJKCR-UHFFFAOYSA-N
1-bromo-4-hexylbenzene
598186
97%
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.