missing translation for 'onlineSavingsMsg'
Learn More

1,4-Butane sultone, 99+%

Product Code. 10234610 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
This item is not returnable. View return policy

Product Code. 10234610

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1633-83-6 | C4H8O3S | 136.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1633-83-6
Molecular Formula C4H8O3S
Molecular Weight (g/mol) 136.17
MDL Number MFCD00006584
InChI Key MHYFEEDKONKGEB-UHFFFAOYSA-N
Synonym 1,4-butane sultone, 1,4-butanesultone, butane sultone, butanesultone, 1,2-oxathiane, 2,2-dioxide, butanesulfone, 1,2-oxathiane 2,2-dioxide, 1,4-butylene sulfone, delta-valerosultone, 1,4-butanesulfone
PubChem CID 15411
IUPAC Name oxathiane 2,2-dioxide
SMILES O=S1(=O)CCCCO1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 12.5°C to 15.0°C
Color Colorless to Yellow
Density 1.3310g/mL
Boiling Point 153°C (19.0 HPA)
Flash Point 154°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4600 to 1.4640
Quantity 100 mL
Beilstein 19, V,1, 10
Specific Gravity 1.331
Solubility Information Solubility in water: 54g/L (20°C), decomposes. Other solubilities: soluble in chloroform
Formula Weight 136.17
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 1, 4-Butane sultone, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,4-Butane sultone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.