missing translation for 'onlineSavingsMsg'
Learn More
1,4-Butanediol diglycidyl ether, 96%, Thermo Scientific Chemicals Product Code.: 11363276

1,4-Butanediol diglycidyl ether, 96%, Thermo Scientific Chemicals

Product Code. 11363276
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11363276

Brand: Thermo Scientific Alfa Aesar L12632.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1,4-Butanediol diglycidyl ether is used for preparing amylose, xylan and hydroxyethyl-cellulose, to cross link polyethylenimine which also serves as a hole-blocking layer in organic solar cells, to cross link hyaluronic acid into hydrogels and for the activation of soluble dextran polymers. It is the most commonly used homobifunctional epoxide compound

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Butanediol diglycidyl ether is used for preparing amylose, xylan and hydroxyethyl-cellulose, to cross link polyethylenimine which also serves as a hole-blocking layer in organic solar cells, to cross link hyaluronic acid into hydrogels and for the activation of soluble dextran polymers. It is the most commonly used homobifunctional epoxide compound

Solubility
Soluble in water (55,6 g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2425-79-8
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00005146
InChI Key SHKUUQIDMUMQQK-UHFFFAOYNA-N
Synonym 1,4-butanediol diglycidyl ether, 1,4-diglycidyloxybutane, 1,4-bis glycidyloxy butane, butanediol diglycidyl ether, araldit dy 026, 1,4-butane diglycidyl ether, grilonit rv 1806, 1,4-diglycidloxybutane, chs-rr2, 1,4-bis 2,3-epoxypropoxy butane
PubChem CID 17046
IUPAC Name 2-[4-(oxiran-2-ylmethoxy)butoxymethyl]oxirane
SMILES C(CCOCC1CO1)COCC1CO1

Specifications

Density 1.049
Boiling Point 157°C to 158°C (11 mmHg)
Flash Point 140°C (284°F)
Refractive Index 1.455
Quantity 50 g
Beilstein 115238
Sensitivity Hygroscopic
Solubility Information Soluble in water (55,6g/L at 20°C).
Formula Weight 202.25
Percent Purity 96%
Chemical Name or Material 1,4-Butanediol diglycidyl ether
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10246402 Certificate TypeCertificate of Analysis Date09/04/2025 Catalog NumberL12632.18,L12632.09
Lot NumberP18L002 Certificate TypeCertificate of Analysis Date20/03/2025 Catalog NumberL12632.18,L12632.09
Lot Number5004Q11W Certificate TypeCertificate of Analysis Date06/12/2024 Catalog NumberL12632.18,L12632.09
Lot Number5001Q04V Certificate TypeCertificate of Analysis Date20/08/2024 Catalog NumberL12632.18,L12632.09
Lot Number10252095 Certificate TypeCertificate of Analysis Date16/08/2024 Catalog NumberL12632.18,L12632.09
compliance-icons compliance-icons
Product Identifier
  • 1,4-Butanediol diglycidyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1,4-Butanediol diglycidyl ether

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,4-Butanediol diglycidyl ether, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.