missing translation for 'onlineSavingsMsg'
Learn More

1,4-Butanesultone, 99%

Product Code. 11316746
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11316746

Brand: Thermo Scientific Alfa Aesar L03992.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1633-83-6 | C4H8O3S | 136.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
1,4-Butanesultone is an alkylating agent used in synthetic chemistry. It serves as a wood preservative. It is employed as an intermediate of the organic sulfonation agent. Further, it is used as a raw material for electroplating intermediate. In addition to this, it finds application in active pharmaceutical ingredients, photographic materials, lithium battery and surface active agents.

Solubility
Miscible with chloroform and methanol. Slightly miscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1633-83-6
Molecular Formula C4H8O3S
Molecular Weight (g/mol) 136.17
MDL Number MFCD00006584
InChI Key MHYFEEDKONKGEB-UHFFFAOYSA-N
Synonym 1,4-butane sultone, 1,4-butanesultone, butane sultone, butanesultone, 1,2-oxathiane, 2,2-dioxide, butanesulfone, 1,2-oxathiane 2,2-dioxide, 1,4-butylene sulfone, delta-valerosultone, 1,4-butanesulfone
PubChem CID 15411
IUPAC Name oxathiane 2,2-dioxide
SMILES O=S1(=O)CCCCO1

Specifications

Melting Point 12°C to 15°C
Density 1.335
Boiling Point 114°C to 116°C (1.5 mmHg)
Flash Point 153°C (307°F)
Refractive Index 1.464
Quantity 25 g
Beilstein 110588
Sensitivity Moisture sensitive
Solubility Information Miscible with chloroform and methanol. Slightly miscible with water.
Formula Weight 136.17
Percent Purity 99%
Chemical Name or Material 1,4-Butanesultone
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.