missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)-1-cyclobutanecarboxylic acid, 94%, Thermo Scientific™

Product Code. 10478140 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10478140

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 50921-39-6
Molecular Formula C11H11ClO2
Molecular Weight (g/mol) 210.66
InChI Key XYSRHOKREWGGFE-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl cyclobutanecarboxylic acid, 1-4-chlorophenyl-1-cyclobutanecarboxylic acid, 1-4-chlorophenyl cyclobutane-1-carboxylic acid, 1-p-chlorophenyl cyclobutanecarboxylic acid, cyclobutanecarboxylic acid, 1-4-chlorophenyl, acmc-209kr1, 1-4chlorophenyl cyclobutanecarboxylic acid, 1 4-chlorophenyl cyclobutane carboxylic acid, 1-4 chlorophenyl-cyclobutanecarboxylic acid, 1-4-chloro phenyl cyclobutanecarboxylic acid
PubChem CID 602761
IUPAC Name 1-(4-chlorophenyl)cyclobutane-1-carboxylic acid
SMILES C1CC(C1)(C2=CC=C(C=C2)Cl)C(=O)O

Specifications

Melting Point 80.0°C to 90.0°C
Color Purple to Yellow
Quantity 25 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Assay Percent Range 92% min. (HPLC)
Linear Formula ClC6H4C4H6CO2H
Formula Weight 210.66
Percent Purity 94%
Chemical Name or Material 1-(4-Chlorophenyl)-1-cyclobutanecarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 1-(4-Chlorophenyl)-1-cyclobutanecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.