missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)-1H-1,2,3-triazole-4-methanol, 98%, Thermo Scientific™

25.00 € - 251.00 €

Chemical Identifiers

CAS 133902-66-6
Molecular Formula C9H8ClN3O
Molecular Weight (g/mol) 209.633
MDL Number MFCD00140928
InChI Key CAHIFLPAMJOAGI-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl-1h-1,2,3-triazol-4-yl methanol, 1-4-chlorophenyl triazol-4-yl methanol, 1-4-chlorophenyl-1,2,3-triazol-4-yl methanol, 1-4-chlorophenyl-1h-1,2,3-triazole-4-yl-methanol, 1-4-chlorophenyl-1,2,3-triazol-4-yl methan-1-ol, pubchem13003, chlorophenyltriazolylmethanol, 1-4-chlorophenyl-1h-1,2,3-triazol-4-ylmethanol
PubChem CID 2763878
IUPAC Name [1-(4-chlorophenyl)triazol-4-yl]methanol
SMILES C1=CC(=CC=C1N2C=C(N=N2)CO)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15489116
View Documents
Thermo Scientific Alfa Aesar
H64737.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15469116
View Documents
Thermo Scientific Alfa Aesar
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15479116
View Documents
Thermo Scientific Alfa Aesar
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

133902-66-6
209.633
CAHIFLPAMJOAGI-UHFFFAOYSA-N
2763878
C1=CC(=CC=C1N2C=C(N=N2)CO)Cl
C9H8ClN3O
MFCD00140928
1-4-chlorophenyl-1h-1,2,3-triazol-4-yl methanol, 1-4-chlorophenyl triazol-4-yl methanol, 1-4-chlorophenyl-1,2,3-triazol-4-yl methanol, 1-4-chlorophenyl-1h-1,2,3-triazole-4-yl-methanol, 1-4-chlorophenyl-1,2,3-triazol-4-yl methan-1-ol, pubchem13003, chlorophenyltriazolylmethanol, 1-4-chlorophenyl-1h-1,2,3-triazol-4-ylmethanol
[1-(4-chlorophenyl)triazol-4-yl]methanol

Specifications

133902-66-6
C9H8ClN3O
4250389
CAHIFLPAMJOAGI-UHFFFAOYSA-N
[1-(4-chlorophenyl)triazol-4-yl]methanol
2763878
98%
1-(4-Chlorophenyl)-1H-1,2,3-triazole-4-methanol
141°C to 145°C
MFCD00140928
1-4-chlorophenyl-1h-1,2,3-triazol-4-yl methanol, 1-4-chlorophenyl triazol-4-yl methanol, 1-4-chlorophenyl-1,2,3-triazol-4-yl methanol, 1-4-chlorophenyl-1h-1,2,3-triazole-4-yl-methanol, 1-4-chlorophenyl-1,2,3-triazol-4-yl methan-1-ol, pubchem13003, chlorophenyltriazolylmethanol, 1-4-chlorophenyl-1h-1,2,3-triazol-4-ylmethanol
C1=CC(=CC=C1N2C=C(N=N2)CO)Cl
209.633
209.63
Solid
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.