missing translation for 'onlineSavingsMsg'
Learn More
1-(4-Chlorophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea, 97%, Thermo Scientific Chemicals Product Code.: 15459616

1-(4-Chlorophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea, 97%, Thermo Scientific Chemicals

Product Code. 15459616
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15459616

Brand: Thermo Scientific Alfa Aesar H65397.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 369-77-7
Molecular Formula C14H9Cl2F3N2O
Molecular Weight (g/mol) 349.13
MDL Number MFCD00867294
InChI Key ZFSXZJXLKAJIGS-UHFFFAOYSA-N
Synonym cloflucarban, halocarban, irgasan cf3, cloflucarbon, trifluoromethyldichlorocarbanilide, unii-i5zzy3dc5g, cloflucarban usan, 4,4'-dichloro-3-trifluoromethyl carbanilide, i5zzy3dc5g, halocarban inn
PubChem CID 9719
IUPAC Name 1-(4-chlorophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea
SMILES FC(F)(F)C1=CC(NC(=O)NC2=CC=C(Cl)C=C2)=CC=C1Cl

Specifications

Quantity 250 mg
Formula Weight 349.13
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(4-Chlorophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(4-Chlorophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.