missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexanediol, 99%, mixture of cis and trans, Thermo Scientific Chemicals

29.04 € - 79.50 €

Chemical Identifiers

CAS 556-48-9
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00001448,MFCD00063612,MFCD00075462
InChI Key VKONPUDBRVKQLM-UHFFFAOYSA-N
Synonym 1,4-cyclohexanediol, cis-1,4-cyclohexanediol, trans-1,4-cyclohexanediol, trans-cyclohexane-1,4-diol, quinitol, 1,4-cyclohexanediol, trans, cis-cyclohexane-1,4-diol, 1,4-cyclohexanediol, cis, trans-1,4-dihydroxycyclohexane, cis-1,4-dihydroxycyclohexane
PubChem CID 11162
IUPAC Name cyclohexane-1,4-diol
SMILES OC1CCC(O)CC1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10343432
View Documents
Thermo Scientific Acros
111150250
25 g Plastic bottle
29.04 €
25g
Please to purchase this item. Need a web account? Register with us today!
10000880
View Documents
Thermo Scientific Acros
111151000
100 g Plastic bottle
79.50 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

556-48-9
116.16
VKONPUDBRVKQLM-UHFFFAOYSA-N
11162
OC1CCC(O)CC1
C6H12O2
MFCD00001448,MFCD00063612,MFCD00075462
1,4-cyclohexanediol, cis-1,4-cyclohexanediol, trans-1,4-cyclohexanediol, trans-cyclohexane-1,4-diol, quinitol, 1,4-cyclohexanediol, trans, cis-cyclohexane-1,4-diol, 1,4-cyclohexanediol, cis, trans-1,4-dihydroxycyclohexane, cis-1,4-dihydroxycyclohexane
cyclohexane-1,4-diol

Specifications

556-48-9
100.0
150°C (20.0 mmHg)
98.5% min. (GC)
C6H10(OH)2
06, IV, 5209
Solubility in water: soluble
OC1CCC(O)CC1
116.16
116.16
1, 4-Cyclohexanediol, 99%
98.5
98°C to 102°C
65°C
C6H12O2
MFCD00001448,MFCD00063612,MFCD00075462
1,4-cyclohexanediol, cis-1,4-cyclohexanediol, trans-1,4-cyclohexanediol, trans-cyclohexane-1,4-diol, quinitol, 1,4-cyclohexanediol, trans, cis-cyclohexane-1,4-diol, 1,4-cyclohexanediol, cis, trans-1,4-dihydroxycyclohexane, cis-1,4-dihydroxycyclohexane
VKONPUDBRVKQLM-UHFFFAOYSA-N
cyclohexane-1,4-diol
11162
99%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 209-126-2

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.