missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexanedione, 98%, Thermo Scientific Chemicals

62.81 € - 664.31 €

Chemical Identifiers

CAS 637-88-7
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00001606
InChI Key DCZFGQYXRKMVFG-UHFFFAOYSA-N
Synonym 1,4-cyclohexanedione, tetrahydroquinone, 1,4-dioxocyclohexane, 1,4-cyclohexandione, 1,4 cyclohexane dione, 1,4-cyclohexane-dione, cyclohexan-1,4-dione, pubchem13686, cyclohexane-1,4-quinone, acmc-1b8ar
PubChem CID 12511
ChEBI CHEBI:28286
IUPAC Name cyclohexane-1,4-dione
SMILES C1CC(=O)CCC1=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11488973
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A11100.14
25 g
62.81 €
25g
Please to purchase this item. Need a web account? Register with us today!
11498973
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A11100.22
100 g
182.91 €
100g
Please to purchase this item. Need a web account? Register with us today!
11408983
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A11100.36
500 g
664.31 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1,4-Cyclohexanedione is used in the preparation of 1,4 benzoquinone and bromoorganics. It is also used to study the influence of visible light on the bromate-1,4-cyclohexanedione-ferroin oscillating reaction. It plays a vital role in pharmaceuticals, plant growth regulator and as a conducting material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Cyclohexanedione is used in the preparation of 1,4 benzoquinone and bromoorganics. It is also used to study the influence of visible light on the bromate-1,4-cyclohexanedione-ferroin oscillating reaction. It plays a vital role in pharmaceuticals, plant growth regulator and as a conducting material.

Solubility
Soluble in methanol and ethanol.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

637-88-7
112.128
DCZFGQYXRKMVFG-UHFFFAOYSA-N
12511
cyclohexane-1,4-dione
C6H8O2
MFCD00001606
1,4-cyclohexanedione, tetrahydroquinone, 1,4-dioxocyclohexane, 1,4-cyclohexandione, 1,4 cyclohexane dione, 1,4-cyclohexane-dione, cyclohexan-1,4-dione, pubchem13686, cyclohexane-1,4-quinone, acmc-1b8ar
CHEBI:28286
C1CC(=O)CCC1=O

Specifications

637-88-7
132°C (20 mmHg)
C6H8O2
774152
Soluble in methanol and ethanol.
C1CC(=O)CCC1=O
112.128
CHEBI:28286
98%
77°C to 78°C
132°C (269°F)
MFCD00001606
1,4-cyclohexanedione, tetrahydroquinone, 1,4-dioxocyclohexane, 1,4-cyclohexandione, 1,4 cyclohexane dione, 1,4-cyclohexane-dione, cyclohexan-1,4-dione, pubchem13686, cyclohexane-1,4-quinone, acmc-1b8ar
DCZFGQYXRKMVFG-UHFFFAOYSA-N
cyclohexane-1,4-dione
12511
112.13
1,4-Cyclohexanedione
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 211-306-0

missing translation for 'tsca' : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only