missing translation for 'onlineSavingsMsg'
Learn More
1,4-Diaminobutane, 99%, Thermo Scientific Chemicals Product Code.: 10386410

1,4-Diaminobutane, 99%, Thermo Scientific Chemicals

Product Code. 10386410
25 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10386410

Brand: Thermo Scientific Acros 112120250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-60-1
Molecular Formula C4H12N2
Molecular Weight (g/mol) 88.15
MDL Number MFCD00008235
InChI Key KIDHWZJUCRJVML-UHFFFAOYSA-N
Synonym 1,4-diaminobutane, putrescine, 1,4-butanediamine, tetramethylenediamine, butylenediamine, putrescin, 1,4-butylenediamine, tetramethyldiamine, 1,4-tetramethylenediamine, putrescina
PubChem CID 1045
ChEBI CHEBI:17148
IUPAC Name butane-1,4-diamine
SMILES C(CCN)CN

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 27.0°C
Color Colorless to Yellow
Density 0.8770g/mL
Boiling Point 158.0°C to 160.0°C
Flash Point 45°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2N(CH2)4NH2
Refractive Index 1.4560 to 1.4610
Quantity 25 g
Beilstein 04,264
Merck Index 15,8058
Specific Gravity 0.877
Solubility Information Solubility in water: 40g/L (20°C).
Formula Weight 88.15
Percent Purity 99%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 1, 4-Diaminobutane, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0469304 Certificate TypeCertificate of Analysis Date23/01/2025 Catalog Number112120025,112121000,112125000,112120250
Lot NumberA0463624 Certificate TypeCertificate of Analysis Date13/06/2024 Catalog Number112120025,112121000,112120250,112125000
Lot NumberA0425407 Certificate TypeCertificate of Analysis Date18/12/2023 Catalog Number112120025,112121000,112120250,112125000
Lot NumberA0440364 Certificate TypeCertificate of Analysis Date13/10/2023 Catalog Number112120025,112121000,112120250,112125000
Lot NumberA0438586 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number112120025,112121000,112120250,112125000
compliance-icons compliance-icons
Product Identifier
  • 1,4-Diaminobutane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H311-Toxic in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,4-Diaminobutane, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.