missing translation for 'onlineSavingsMsg'
Learn More
1,4-Diazabicyclo[2.2.2]octane, 97%, Thermo Scientific Chemicals Product Code.: 10214620

1,4-Diazabicyclo[2.2.2]octane, 97%, Thermo Scientific Chemicals

Product Code. 10214620
100 g, Glass Bottle
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10214620

Brand: Thermo Scientific Acros 112471000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 280-57-9
Molecular Formula C6H12N2
Molecular Weight (g/mol) 112.17
MDL Number MFCD00006689
InChI Key IMNIMPAHZVJRPE-UHFFFAOYSA-N
Synonym 1,4-diazabicyclo 2.2.2 octane, triethylenediamine, dabco, dabco 33lv, 1,4-ethylenepiperazine, dabco crystal, teda, texacat td 100, dabco s-25
PubChem CID 9237
IUPAC Name 1,4-diazabicyclo[2.2.2]octane
SMILES C1CN2CCN1CC2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 155.0°C to 160.0°C
Color White to Yellow
Density 1.1400g/mL
Boiling Point 174.0°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 100 g
Fieser 01,1203; 02,99; 04,119; 05,176; 06,157; 07,86; 11,153; 12,155; 13,92; 15,109
Merck Index 15,9834
Specific Gravity 1.14
Solubility Information Solubility in water: 46g/100mL (26°C). Other solubilities: soluble in most common organic solvents
Sublimation Point 20 °C
Formula Weight 112.17
Percent Purity 97%
Physical Form Hygroscopic Crystals
Chemical Name or Material 1,4-Diazabicyclo[2.2.2]octane, 97%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,4-Diazabicyclo¢2.2.2!octane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,4-Diazabicyclo[2.2.2]octane, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.