missing translation for 'onlineSavingsMsg'
Learn More
1,4-Diazabicyclo[2.2.2]octane, 98%, Thermo Scientific Chemicals Product Code.: 11407866

1,4-Diazabicyclo[2.2.2]octane, 98%, Thermo Scientific Chemicals

Product Code. 11407866
500 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11407866

Brand: Thermo Scientific Alfa Aesar A14003.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1,4-Diazabicyclo[2.2.2]octane is used as polyurethane catalyst, Balis-Hillman reaction catalyst complexing ligand and lewis base. It finds use in dye lasers and in mounting samples for fluorescence microscopy and as anti-fade reagent shown to scavenge free radicals due to flurochrome excitation of fluorochromes. Further, it is an oxidation and polymerization catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Diazabicyclo[2.2.2]octane is used as polyurethane catalyst, Balis-Hillman reaction catalyst complexing ligand and lewis base. It finds use in dye lasers and in mounting samples for fluorescence microscopy and as anti-fade reagent shown to scavenge free radicals due to flurochrome excitation of fluorochromes. Further, it is an oxidation and polymerization catalyst.

Solubility
Soluble in water, acetone, benzene, ethanol and methyl ethyl ketone.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 280-57-9
Molecular Formula C6H12N2
Molecular Weight (g/mol) 112.176
MDL Number MFCD00006689
InChI Key IMNIMPAHZVJRPE-UHFFFAOYSA-N
Synonym 1,4-diazabicyclo 2.2.2 octane, triethylenediamine, dabco, dabco 33lv, 1,4-ethylenepiperazine, dabco crystal, teda, texacat td 100, dabco s-25
PubChem CID 9237
IUPAC Name 1,4-diazabicyclo[2.2.2]octane
SMILES C1CN2CCN1CC2

Specifications

Melting Point 155°C to 160°C
Density 1.14
Boiling Point 174°C to 176°C
Flash Point 62°C (143°F)
Odor Amine-like
Quantity 500 g
UN Number UN1325
Beilstein 103618
Sensitivity Hygroscopic
Merck Index 14,9669
Solubility Information Soluble in water,acetone,benzene,ethanol and methyl ethyl ketone.
Formula Weight 112.17
Percent Purity 98%
Chemical Name or Material 1,4-Diazabicyclo[2.2.2]octane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number5002Q18S Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberA14003.36
Lot Number10251982 Certificate TypeCertificate of Analysis Date12/08/2024 Catalog NumberA14003.36,A14003.22
Lot Number10246998 Certificate TypeCertificate of Analysis Date20/06/2023 Catalog NumberA14003.36,A14003,A14003.22
Lot NumberU26I001 Certificate TypeCertificate of Analysis Date22/09/2022 Catalog NumberA14003.36,A14003.22
Lot NumberP31I042 Certificate TypeCertificate of Analysis Date14/04/2022 Catalog NumberA14003.36,A14003.22
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,4-Diazabicyclo[2.2.2]octane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H228-Flammable solid.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 1,4-Diazabicyclo[2.2.2]octane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,4-Diazabicyclo[2.2.2]octane, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.