missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromobutane, 98+%, Thermo Scientific Chemicals

24.62 € - 73.16 €

Chemical Identifiers

CAS 110-52-1
Molecular Formula C4H8Br2
Molecular Weight (g/mol) 215.916
MDL Number MFCD00000261
InChI Key ULTHEAFYOOPTTB-UHFFFAOYSA-N
Synonym tetramethylene dibromide, butane, 1,4-dibromo, 1,4-butylene bromide, tetramethylene bromide, 1,4-dibrombutan, tetramethylenebromide, alpha,omega-dibromobutane, unii-g49phr6jfu, 1,4-dibrombutan german, 1,4-dibromo-butane
PubChem CID 8056
IUPAC Name 1,4-dibromobutane
SMILES C(CCBr)CBr
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11424220
View Documents
Thermo Scientific Alfa Aesar
A11669.09
10 g
24.62 €
10g
Please to purchase this item. Need a web account? Register with us today!
11419153
View Documents
Thermo Scientific Alfa Aesar
A11669.22
100 g
34.07 €
100g
Please to purchase this item. Need a web account? Register with us today!
11429153
View Documents
Thermo Scientific Alfa Aesar
A11669.36
500 g
73.16 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1,4-Dibromobutane is used as an intermediate involved in the synthesis of active pharmaceutical ingredient and other organic compounds. It is also used in the investigation of metabolism of two halopropanes such as 1,3-dichloropropane and 2,2-dichloropropane. Further, it acts as a reagent to prepare diazadioxa oxovanadium(IV) macrocyclic complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dibromobutane is used as an intermediate involved in the synthesis of active pharmaceutical ingredient and other organic compounds. It is also used in the investigation of metabolism of two halopropanes such as 1,3-dichloropropane and 2,2-dichloropropane. Further, it acts as a reagent to prepare diazadioxa oxovanadium(IV) macrocyclic complexes.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications

Chemical Identifiers

110-52-1
215.916
ULTHEAFYOOPTTB-UHFFFAOYSA-N
8056
C(CCBr)CBr
C4H8Br2
MFCD00000261
tetramethylene dibromide, butane, 1,4-dibromo, 1,4-butylene bromide, tetramethylene bromide, 1,4-dibrombutan, tetramethylenebromide, alpha,omega-dibromobutane, unii-g49phr6jfu, 1,4-dibrombutan german, 1,4-dibromo-butane
1,4-dibromobutane

Specifications

110-52-1
1.82
131°C (267°F)
Br(CH2)4Br
MFCD00000261
tetramethylene dibromide, butane, 1,4-dibromo, 1,4-butylene bromide, tetramethylene bromide, 1,4-dibrombutan, tetramethylenebromide, alpha,omega-dibromobutane, unii-g49phr6jfu, 1,4-dibrombutan german, 1,4-dibromo-butane
ULTHEAFYOOPTTB-UHFFFAOYSA-N
1,4-dibromobutane
8056
≥98%
-20°C
194°C to 196°C
C4H8Br2
1.5186
1071199
Immiscible with water.
C(CCBr)CBr
215.916
215.93
1,4-Dibromobutane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H301-H315-H318-H335

missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III

missing translation for 'einecsNumber' : 203-775-5

missing translation for 'rtecsNumber' : EJ7565000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.